Learn More
Invitrogen™ ATP Solution (10 mM)
This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits and is supplied in one tube containing 100 μL.
Supplier: Invitrogen™ AM8110G
Description
This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308) and is supplied in one tube containing 100 μL. Each is subjected to RNase testing as well as a functional assay to ensure high levels of incorporation. It may also be used in ligation and phosphorylation reactions.
Order Info
Shipping Condition: Dry Ice
Specifications
| Store at -20°C. | |
| In Vitro transcription | |
| Unlabeled | |
| 100 μL |
| 10 mM | |
| RNA Labeling | |
| Ambion | |
| Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.