missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ArrayControl™ RNA Spikes

Catalog No. AM1780
Encompass
Change view
Click to view available options
Quantity:
8 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1780 8 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1780 Supplier Invitrogen™ Supplier No. AM1780

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Add to the RNA sample for use as a template for preparing labeled cDNA

When hybridized to complementary spots on a gene array, ArrayControl RNA Spikes can be used to monitor the efficiency of reverse transcription and aRNA amplification, labeling, and hybridization. They can also be used to assess the precision, accuracy, and consistency of array printing and protocol sensitivity. In addition, RNA Spikes are an invaluable tool for protocol optimization and troubleshooting. They can be used for normalization of Cy3 and Cy5 signals, determination of dynamic range, and slide orientation. Individual RNA Spikes can also be omitted to serve as a negative control for background calculations. The RNA Spike sequences have been carefully chosen to lack homology to mammalian sequences that might otherwise lead to cross-hybridization to gene-specific spots. Each RNA Spike is purified, precisely quantitated by spectrophotometry and RiboGreen™ staining, and tested for stability under highly stringent conditions. They are also tested for cDNA labeling efficiency using both direct and indirect methods. Their manufacturing and quantitation are consistent from lot-to-lot, so that signals can be compared between experiments and to assess array reproducibility.

  • Consists of 8 individual RNA transcripts ranging in size from 750 to 2000 bases with a 30-base 3' poly(A) tail
  • Each transcript is supplied in one tube containing 1μg
  • For the normalization and control of microarray experiments
  • For array printing and protocol assessment/optimization
  • Monitor reverse transcription and cDNA labeling efficiency, hybridization stringency, and background
  • Normalize Cy™3 and Cy5 signals

Gene Expression Analysis and Genotyping, Microarray Analysis

Order Info

Shipping Condition: Dry Ice

Specifications

Product Type RNA Spike
Content And Storage RNA Spikes 1–8 should be stored below –70°C. TE Buffer should be stored at –20°C.
Label or Dye Unconjugated
Shipping Condition Dry Ice
Product Line Ambion, ArrayControl
Quantity 8 Tubes

For Research Use Only. Not for use in diagnostic procedures.