missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amberlyst™ A26 Hydroxide Form, MilliporeSigma™ Supelco™
Inert in strong acids, concentrated aldalies, aliphatic and aromatic hydrocarbons, alcohols, ethers, and other common solvents.Maximum operating temperature 60C.
$141.10 - $699.46
Specifications
| CAS | 39339-85-0 |
|---|---|
| Product Line | Amberlyst |
| Physical Form | Beads |
| Concentration | 1 N in NaOH (Regenerant concentration) |
| MDL Number | MFCD03096474 |
Specifications
| 39339-85-0 | |
| Beads | |
| MFCD03096474 |
| Amberlyst | |
| 1 N in NaOH (Regenerant concentration) | |
| Organic Standard |
Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow