missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ A26 Hydroxide Form, MilliporeSigma™ Supelco™

Catalog No. 111014112
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014112 100 g
111014113 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014112 Supplier Merck Emd Millipore Supplier No. 542571100G

Inert in strong acids, concentrated aldalies, aliphatic and aromatic hydrocarbons, alcohols, ethers, and other common solvents.Maximum operating temperature 60C.

Specifications

CAS 39339-85-0
Product Line Amberlyst
Physical Form Beads
Concentration 1 N in NaOH (Regenerant concentration)
MDL Number MFCD03096474
Type Organic Standard
Quantity 100 g

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow