missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ AmberLite HPR4800 Cl, Chloride Form
Formerly Dowex™ Marathon™ A, chloride form
$193.96 - $566.01
Specifications
| Packaging | Poly Bottle |
|---|---|
| Product Line | Marathon |
| Capacity (Metric) | 1.3 meq/mL by wetted bed volume |
| MDL Number | MFCD00145586 |
| CAS | 60177-39-1 |
Description
Dowex Marathon A is a strong basic anionic Type I exchange gel resin with trimethylamine as functional group. Dowex Marathon A is a heterogeneous catalyst made of styerenedivinylbenzene copolymer.Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow