missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ AmberLite HPR4800 Cl, Chloride Form

Catalog No. 111014832
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014832 250 g
111014831 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014832 Supplier MilliporeSigma™ Supelco™ Supplier No. 433942250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formerly Dowex™ Marathon™ A, chloride form

Dowex Marathon A is a strong basic anionic Type I exchange gel resin with trimethylamine as functional group. Dowex Marathon A is a heterogeneous catalyst made of styerenedivinylbenzene copolymer.

Specifications

Packaging Poly Bottle
Quantity 250 g
Product Line Marathon
Capacity (Metric) 1.3 meq/mL by wetted bed volume
MDL Number MFCD00145586
CAS 60177-39-1

Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow