Learn More
MilliporeSigma™ Supelco™ AmberLite HPR4800 Cl, Chloride Form
Formerly Dowex™ Marathon™ A, chloride form
Supplier:  MilliporeSigma™ Supelco™ 4339421KG
 View more versions of this product
Catalog No. 111014831
Only null left
Explore available promotions
Description
Description
Dowex Marathon A is a strong basic anionic Type I exchange gel resin with trimethylamine as functional group. Dowex Marathon A is a heterogeneous catalyst made of styerenedivinylbenzene copolymer.
Specifications
Specifications
Poly Bottle
Marathon
MFCD00145586
1 kg
1.3 meq/mL by wetted bed volume
60177-39-1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow