Learn More
Fisherbrand™ Amber Wide Mouth Packer Glass Bottles without Caps
Amber, USP Type III soda-lime glass ideal for storage of light sensitive samples or dry powders. Easy removal of contents.
 View more versions of this product
Catalog No. FB02911888
Only null left
Description
Description
  • Features a wide opening
  • Amber color filters out ultraviolet wavelength
  • Often used in chemical and environmental applications
Specifications
Chemical Identifiers
Amber
No
Wide Mouth Packer Bottle
Glass (Type 3)
Wide
Specifications
120 mL
35 mm
Glass (Type 3)
38-400
Wide
Wide Mouth Packer Bottle
54 mm
Amber
25 mm
180
No
95 mm
38-400 mm
Product Suggestions
Product Suggestions