missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Amber Reagent Bottle , Narrow Mouth with Acid Proof Polypropylene Stopper
Narrow Mouth with Acid Proof Polypropylene Stopper. Made of high quality borosilicate glass.
$59.11 - $383.07
Specifications
| Color | Amber |
|---|---|
| For Use With (Application) | To contain chemicals in liquid or powder form |
| Material | Borosilicate Glass |
| Quantity | 1 |
| Sterility | Non-sterile |
Specifications
| Amber | |
| Borosilicate Glass | |
| Non-sterile | |
| Yes | |
| No | |
| Cylindrical | |
| Acid | |
| Polypropylene | |
| Autoclavable |
| To contain chemicals in liquid or powder form | |
| 1 | |
| White | |
| Reusable | |
| Narrow | |
| Reagent Bottle | |
| Unlabeled | |
| Stopper |