missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Amber Reagent Bottle , Narrow Mouth with Acid Proof Polypropylene Stopper

Catalog No. S27979
Change view
Click to view available options
Capacity (Metric):
125 mL
1000 mL
2000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S27979 2000 mL
S27975 125 mL
S27978 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S27979 Supplier Eisco™ Supplier No. CH0161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Narrow Mouth with Acid Proof Polypropylene Stopper. Made of high quality borosilicate glass.

Specifications

Capacity 2000 mL
Capacity (Metric) 2000 mL
Color Amber
Description Socket Size: 34/35
For Use With (Application) To contain chemicals in liquid or powder form
Material Borosilicate Glass
Length (Metric) 330.2 mm
Quantity 1
Sterility Non-sterile
Standard Taper Joint 34/35
Weight (Metric) 907.18 g
Width (Metric) 152.4 mm
Closure Color White
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 15.24 cm
For Use With (Equipment) Acid
Height (English) 6 in.
Chemical Label Unlabeled
Closure Material Polypropylene
Closure Size 34/35
Closure Type Stopper
Autoclavable Autoclavable
Capacity (English) 67.62 oz.
Shape Cylindrical
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less