Learn More
Fisherbrand™ Amber Boston Round Glass Bottles without Caps
Type III soda-lime glass is ideal for solvent, chemical or sample storage
 View more versions of this product
Catalog No. FB02911902
Only null left
Description
Description
  • Features narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
  • Ensure integrity for long term storage and protection for UV-sensitive contents
Specifications
Chemical Identifiers
Amber
No
Boston Round Bottle
Glass (Type 3)
Narrow
Specifications
480 mL
Amber
16 mm
60
No
168 mm
28-400 mm
16 oz.
25 mm
Glass (Type 3)
28-400
Narrow
Boston Round Bottle
74 mm
Product Suggestions
Product Suggestions