missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum sulfate hydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis, Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis
$1289.48 - $1289.48
Chemical Identifiers
| CAS | 7784-31-8 |
|---|---|
| Molecular Formula | Al2H36O30S3 |
| Molecular Weight (g/mol) | 666.401 |
| InChI Key | AMVQGJHFDJVOOB-UHFFFAOYSA-H |
| Synonym | aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 |
| PubChem CID | 22377415 |
| ChEBI | CHEBI:74779 |
| IUPAC Name | dialuminum;trisulfate;octadecahydrate |
| SMILES | O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7784-31-8 | |
| 666.401 | |
| aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
| CHEBI:74779 | |
| O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
| Al2H36O30S3 | |
| AMVQGJHFDJVOOB-UHFFFAOYSA-H | |
| 22377415 | |
| dialuminum;trisulfate;octadecahydrate |
Specifications
| 7784-31-8 | |
| Al2H36O30S3 | |
| UN3260 | |
| AMVQGJHFDJVOOB-UHFFFAOYSA-H | |
| dialuminum;trisulfate;octadecahydrate | |
| 22377415 | |
| 342.15 (anhydrous basis) | |
| puriss. |
| 6 x 1 kg | |
| Al2(SO4)3 · 18H2O | |
| aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
| O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] | |
| 666.401 | |
| CHEBI:74779 | |
| 100 to 110% | |
| Aluminum Sulfate Hydrate, Meets analytical specification of Ph.Eur.BP |
Safety and Handling
P280-P305 + P351 + P338 + P310
H290-H318
DOTInformation : Class 8 - PG 3 - RQ - Corrosive solid, acidic, inorganic, n.o.s., HI: all
EINECSNumber : 233-135-0
RTECSNumber : WS5697000