missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLAL22X

 View more versions of this product

Catalog No. PLAL2-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Aluminum Standard
7429-90-5,7732-18-5,7697-37-2
Al
AA, ICP
Certified Reference Material
[Al]
aluminium
500 mL
CHEBI:33629
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00134029
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
XAGFODPZIPBFFR-UHFFFAOYSA-N
2% HNO3
26.98
5359268
Assurance

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
CHEBI:33629
[Al]
Al
MFCD00134029
5359268
aluminium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only