missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$274.88 - $549.79

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLAL2-2Y
View Documents
SPEX CertiPrep Group LLC
PLAL22Y
125 mL Triple-leached LDPE bottle
Each for $274.88
Only null left
 
PLAL22T
View Documents
SPEX CertiPrep Group LLC
PLAL22T
250 mL Triple-leached LDPE bottle
Each for $410.56
Only null left
 
PLAL2-2X
View Documents
SPEX CertiPrep Group LLC
PLAL22X
500 mL Triple-leached LDPE bottle
Each for $549.79
Only null left
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
CHEBI:33629
[Al]
Al
MFCD00134029
5359268
aluminium

Specifications

Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00134029
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
XAGFODPZIPBFFR-UHFFFAOYSA-N
2% HNO3
26.98
5359268
Assurance
7429-90-5,7732-18-5,7697-37-2
Al
AA, ICP
Certified Reference Material
[Al]
aluminium
125 mL
CHEBI:33629
Aluminum Standard
Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only