missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
CAS: 7429-90-5 | Al | 26.98 g/mol
$98.01 - $332.52
Chemical Identifiers
| CAS | 7429-90-5 |
|---|---|
| Molecular Formula | Al |
| Molecular Weight (g/mol) | 26.98 |
| MDL Number | MFCD00134029 |
| InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
| PubChem CID | 5359268 |
| ChEBI | CHEBI:33629 |
| SMILES | [Al] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA13856AD
|
Thermo Scientific Chemicals
013856AD |
50 mL |
Each for $98.01
|
|
|||||
|
AA13856AE
|
Thermo Scientific Chemicals
013856AE |
100 mL |
Each for $139.99
|
|
|||||
|
AA13856AP
|
Thermo Scientific Chemicals
013856AP |
500 mL |
Each for $332.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7429-90-5 | |
| 26.98 | |
| XAGFODPZIPBFFR-UHFFFAOYSA-N | |
| CHEBI:33629 |
| Al | |
| MFCD00134029 | |
| 5359268 | |
| [Al] |
Specifications
| 7429-90-5 , 7732-18-5 | |
| Liquid | |
| MFCD00134029 | |
| [Al] | |
| 26.98 | |
| 5359268 | |
| Hydrochloric acid: 5.0%; Aluminum: 0.1%; Water: 94.9% |
| 23 hPa (17mm Hg) at 20°C | |
| Al | |
| XAGFODPZIPBFFR-UHFFFAOYSA-N | |
| aluminium | |
| 50 mL | |
| CHEBI:33629 | |
| Aluminum |
Safety and Handling
P234-P390
H290-H335
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 231-595-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only