Learn More
Aluminum Monostearate, NF, 14.7-16.7%, Spectrum™ Chemical
AL228, 7047-84-9
$1366.73 - $138860.90
Chemical Identifiers
CAS 7047-84-9
View More Specs
Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600479
View Documents
Spectrum Chemical Mfg Cor
AL228100GM
100 g Poly Bottle
Each for $1,366.73
Only null left
18600480
View Documents
Spectrum Chemical Mfg Cor
AL228500GM
500 g Poly Pail
Each for $4,705.83
Only null left
18600481
View Documents
Spectrum Chemical Mfg Cor
AL22825KGBL
2.5 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18600482
View Documents
Spectrum Chemical Mfg Cor
AL22812KGBL
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18600483
View Documents
Spectrum Chemical Mfg Cor
AL22850KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Spectrum™ Chemical Aluminum Monostearate, NF , also known as Dihydroxyaluminum stearate, is used to form pharmaceutical gels. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Chemical Identifiers
7047-84-9
Specifications
7047-84-9
100 g
NF
Poly Bottle
100%
14.7 to 16.7%
3%
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'