missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Monostearate, NF, 14.7-16.7%, Spectrum™ Chemical
Description
Spectrum™ Chemical Aluminum Monostearate, NF , also known as Dihydroxyaluminum stearate, is used to form pharmaceutical gels. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 50 kg |
| Percent Purity | 14.7 to 16.7% |
| Grade | NF |
| Loss on Drying | 3% |
| Packaging | Fiber Drum |