missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Monostearate, NF, 14.7-16.7%, Spectrum™ Chemical

Catalog No. 18600479
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600479 100 g Poly Bottle
18600480 500 g Poly Pail
18600481 2.5 kg Poly Pail
18600482 12 kg Poly Pail
18600483 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600479 Supplier Spectrum Chemical Mfg Cor Supplier No. AL228100GM
Only null left

AL228, 7047-84-9

Spectrum™ Chemical Aluminum Monostearate, NF , also known as Dihydroxyaluminum stearate, is used to form pharmaceutical gels. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7047-84-9

Specifications

CAS Max % 100%
Quantity 100 g
Percent Purity 14.7 to 16.7%
Grade NF
Loss on Drying 3%
Packaging Poly Bottle