missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride Solution, For Sulfide, 6N, LabChem™

Catalog No. p-5388350
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC108207 125 mL
LC108201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC108207 Supplier LabChem Supplier No. LC108207
Only null left

Chemical Identifiers

CAS 7784-13-6 , 7732-18-5

Specifications

Color Colorless to Yellow
CAS Max % 50
Molecular Formula ClH4N
Linear Formula AlCl3·6H2O
MDL Number MFCD00011420
UN Number UN2581
Concentration 6N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
Solubility Information Soluble in water
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC Name amine hydrochloride
Molecular Weight (g/mol) 53.49
PubChem CID 25517
ChEBI CHEBI:31206
Formula Weight 241.43
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Aluminum Chloride Solution, For Sulfide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing mist.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature