missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina, Activated, 80-200 Mesh, Chromatographic Grade, Spectrum™ Chemical
AL219, 1344-28-1, Al2O3
$585.73 - $18845.47
Chemical Identifiers
| CAS | 1344-28-1 |
|---|---|
| Molecular Formula | Al2O3 |
| Molecular Weight (g/mol) | 101.96 |
| MDL Number | MFCD00003424 |
| InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
| IUPAC Name | dialuminium(3+) trioxidandiide |
| SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600456
|
Spectrum Chemical Mfg Cor
AL219500GM |
500 g | Amber Glass Bottle |
Each for $585.73
|
|
||||
|
18600457
|
Spectrum Chemical Mfg Cor
AL21925KG |
2.5 kg | Amber Glass Bottle |
Each for $1,834.37
|
|
||||
|
18600459
|
Spectrum Chemical Mfg Cor
AL21945KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Alumina, Activated, 80-200 Mesh, Chromatographic Grade is used for a wide range of adsorbent and catalyst applications. Alumina (aluminum oxides) are suitable for column chromatography and adsorptive filtration. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
| Al2O3 | |
| MFCD00003424 | |
| dialuminium(3+) trioxidandiide |
Specifications
| 1344-28-1 | |
| 500 g | |
| MFCD00003424 | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| Amber Glass Bottle |
| 100% | |
| Al2O3 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminium(3+) trioxidandiide | |
| Chromatography |