missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina, Activated, 80-200 Mesh, Chromatographic Grade, Spectrum™ Chemical
AL219, 1344-28-1, Al2O3
Supplier: Spectrum Chemical Mfg Cor AL21945KGBL
Description
Spectrum™ Chemical Alumina, Activated, 80-200 Mesh, Chromatographic Grade is used for a wide range of adsorbent and catalyst applications. Alumina (aluminum oxides) are suitable for column chromatography and adsorptive filtration. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 1344-28-1 | |
| 45 kg | |
| MFCD00003424 | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| Fiber Drum |
| 100% | |
| Al2O3 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminium(3+) trioxidandiide | |
| Chromatography |
Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
| Al2O3 | |
| MFCD00003424 | |
| dialuminium(3+) trioxidandiide |