missing translation for 'onlineSavingsMsg'
Learn More

Alumina, Activated, 80-200 Mesh, Chromatographic Grade, Spectrum™ Chemical

Catalog No. 18600456
Change view
Click to view available options
Quantity:
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600456 500 g Amber Glass Bottle
18600457 2.5 kg Amber Glass Bottle
18600459 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600456 Supplier Spectrum Chemical Mfg Cor Supplier No. AL219500GM
Only null left

AL219, 1344-28-1, Al2O3

Spectrum™ Chemical Alumina, Activated, 80-200 Mesh, Chromatographic Grade is used for a wide range of adsorbent and catalyst applications. Alumina (aluminum oxides) are suitable for column chromatography and adsorptive filtration. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

CAS Max % 100%
Quantity 500 g
Grade Chromatography
Packaging Amber Glass Bottle