missing translation for 'onlineSavingsMsg'
Learn More

Alumina A, Activity: Super I, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™

MP EcoChrom™ Alumina A - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.

$352.80 - $8843.31

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN04595
View Documents
MP Biomedicals Inc
0204595
1 kg
Each for $352.80
Only null left
 
ICN04598
View Documents
MP Biomedicals Inc
0204598
5 kg
Each for $1,221.73
Only null left
 
ICN04601
View Documents
MP Biomedicals Inc
0204601
50 kg
Each for $8,843.31
Only null left
 
Description

Description

  • Adsorption capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
  • Inital activity is kept in a very narrow range.
  • Extremely constant deactivation behavior.
  • Commonly used for purification processes requiring the highest precision and reproducibility.
Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Pink to White
Free-flowing Powder
Al2O3
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Alumina A, Activity: Super I
1344-28-1
1 kg
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
3.94 g/cm3 (at 20°C)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

Recommended Storage : Store at room temperature.

SDS
missing translation for 'documents'

missing translation for 'documents'

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.