missing translation for 'onlineSavingsMsg'
Learn More

Alumina A, Activity: Super I, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™

MP EcoChrom™ Alumina A - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.

Supplier:  MP Biomedicals Inc 0204595

 View more versions of this product

Catalog No. ICN04595


Only null left
Explore available promotions

Description

Description

  • Adsorption capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
  • Inital activity is kept in a very narrow range.
  • Extremely constant deactivation behavior.
  • Commonly used for purification processes requiring the highest precision and reproducibility.
Specifications

Specifications

Alumina A
Pink to White
Free-flowing Powder
Al2O3
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Activity: Super I
1344-28-1
1 kg
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
3.94 g/cm3 (at 20°C)

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.