Learn More
Alumina A, Activity: Super I, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™
MP EcoChrom™ Alumina A - Super I is commonly used for purification processes of all kinds including sample preparation for pesticides environmental analysis, solvent purification and all kinds of chromatography requiring the highest precision and reproducibility.
Supplier: MP Biomedicals Inc 0204598
Description
- Adsorption capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
- Inital activity is kept in a very narrow range.
- Extremely constant deactivation behavior.
- Commonly used for purification processes requiring the highest precision and reproducibility.
Specifications
| Alumina A | |
| Pink to White | |
| Free-flowing Powder | |
| Al2O3 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| 101.96 |
| Activity: Super I | |
| 1344-28-1 | |
| 5 kg | |
| MFCD00003424 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminium(3+) trioxidandiide | |
| 9989226 | |
| 3.94 g/cm3 (at 20°C) |
Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| 9989226 |
| Al2O3 | |
| MFCD00003424 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
Safety and Handling
EINECSNumber : 215-691-6
Recommended Storage : Store at room temperature.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.