missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Terpinene 90.0+%, TCI America™
$49.36 - $284.79
Chemical Identifiers
| CAS | 99-86-5 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00001534 |
| InChI Key | YHQGMYUVUMAZJR-UHFFFAOYSA-N |
| Synonym | alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene |
| PubChem CID | 7462 |
| ChEBI | CHEBI:10334 |
| IUPAC Name | 1-methyl-4-(propan-2-yl)cyclohexa-1,3-diene |
| SMILES | CC(C)C1=CC=C(C)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
M031725ML
|
N/A | 25 mL |
Each for $49.36
|
|
|||||
|
M0317500ML
|
N/A | 500 mL |
Each for $284.79
|
|
|||||
Chemical Identifiers
| C10H16 | |
| MFCD00001534 | |
| alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene | |
| CHEBI:10334 | |
| CC(C)C1=CC=C(C)CC1 |
Specifications
| 99-86-5 | |
| 173°C | |
| MFCD00001534 | |
| alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene | |
| CC(C)C1=CC=C(C)CC1 | |
| 136.24 | |
| CHEBI:10334 | |
| ≥90.0% (GC) | |
| alpha-Terpinene |
| Yellow | |
| C10H16 | |
| 2319 | |
| YHQGMYUVUMAZJR-UHFFFAOYSA-N | |
| 1-methyl-4-(propan-2-yl)cyclohexa-1,3-diene | |
| 7462 | |
| 136.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2244
RTECSNumber : OS8060000
TSCA : Yes
Recommended Storage : Refrigerator