missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-alpha-Phellandrene 65.0+%, TCI America™
Supplier: TCI America M005125ML
| Quantity | 25 mL |
|---|
Chemical Identifiers
| C10H16 | |
| MFCD00062988 | |
| (-)-5-Isopropyl-2-methyl-1,3-cyclohexadiene, (-)-p-Mentha-1,5-diene | |
| CHEBI:301 | |
| CC1=CCC(C=C1)C(C)C |
Specifications
| (-)-alpha-Phellandrene | |
| Yellow | |
| C10H16 | |
| MFCD00062988 | |
| (-)-5-Isopropyl-2-methyl-1,3-cyclohexadiene, (-)-p-Mentha-1,5-diene | |
| CC1=CCC(C=C1)C(C)C | |
| 136.238 | |
| CHEBI:301 | |
| ≥65.0% (GC) |
| 4221-98-1 | |
| 175°C | |
| 25 mL | |
| 2319 | |
| OGLDWXZKYODSOB-SNVBAGLBSA-N | |
| (5R)-2-methyl-5-propan-2-ylcyclohexa-1,3-diene | |
| 442482 | |
| 136.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2243
TSCA : Yes
Recommended Storage : Refrigerator