missing translation for 'onlineSavingsMsg'
Learn More

(-)-alpha-Phellandrene 65.0+%, TCI America™

Catalog No. p-7131952
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M00515ML 5 mL
M005125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M00515ML Supplier TCI America Supplier No. M00515ML
Only null left

Chemical Identifiers

CAS 4221-98-1
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00062988
InChI Key OGLDWXZKYODSOB-SNVBAGLBSA-N
Synonym (-)-5-Isopropyl-2-methyl-1,3-cyclohexadiene, (-)-p-Mentha-1,5-diene
PubChem CID 442482
ChEBI CHEBI:301
IUPAC Name (5R)-2-methyl-5-propan-2-ylcyclohexa-1,3-diene
SMILES CC1=CCC(C=C1)C(C)C

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 mL
UN Number 2319
Formula Weight 136.24
Percent Purity ≥65.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-alpha-Phellandrene
EINECSNumber (3)-2243
TSCA Yes
Recommended Storage Refrigerator