accessibility menu, dialog, popup

UserName

CAS RN 4221-98-1

H3CCH3(R)CH3

CAS RN 4221-98-1

IUPAC Name: (5R)-2-methyl-5-(propan-2-yl)cyclohexa-1,3-diene
Synonyms: (-)-α-phellandrene α-phellandrene l-form (R)-(-)-α-phellandrene isopropyl-methylcyclohexadiene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: OGLDWXZKYODSOB-SNVBAGLBSA-N
SMILES: CC(C)[C@H]1CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results