missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Naphtholphthalein 98.0+%, TCI America™
$61.51 - $175.22
Chemical Identifiers
| CAS | 596-01-0 |
|---|---|
| Molecular Formula | C28H18O4 |
| Molecular Weight (g/mol) | 418.448 |
| MDL Number | MFCD00036202 |
| InChI Key | HQHBAGKIEAOSNM-UHFFFAOYSA-N |
| Synonym | alpha-naphtholphthalein, naphtholphthalein, 1-naphtholphthalein, a-naphtholphthalein, 3,3-bis 4-hydroxynaphthalen-1-yl isobenzofuran-1 3h-one, unii-2qeg1706nn, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-1-naphthalenyl, naphtholphthalein blue, alpha-naphtholphtalein, 3,3-bis 4-hydroxy-1-naphthyl phthalide |
| PubChem CID | 68993 |
| IUPAC Name | 3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one |
| SMILES | C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O |
Chemical Identifiers
| 596-01-0 | |
| 418.448 | |
| HQHBAGKIEAOSNM-UHFFFAOYSA-N | |
| 68993 | |
| C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O |
| C28H18O4 | |
| MFCD00036202 | |
| alpha-naphtholphthalein, naphtholphthalein, 1-naphtholphthalein, a-naphtholphthalein, 3,3-bis 4-hydroxynaphthalen-1-yl isobenzofuran-1 3h-one, unii-2qeg1706nn, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-1-naphthalenyl, naphtholphthalein blue, alpha-naphtholphtalein, 3,3-bis 4-hydroxy-1-naphthyl phthalide | |
| 3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one |
Specifications
| 596-01-0 | |
| C28H18O4 | |
| 1 g | |
| HQHBAGKIEAOSNM-UHFFFAOYSA-N | |
| 3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one | |
| 68993 | |
| ≥98.0% (HPLC) | |
| alpha-Naphtholphthalein |
| 240°C | |
| MFCD00036202 | |
| alpha-naphtholphthalein, naphtholphthalein, 1-naphtholphthalein, a-naphtholphthalein, 3,3-bis 4-hydroxynaphthalen-1-yl isobenzofuran-1 3h-one, unii-2qeg1706nn, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-1-naphthalenyl, naphtholphthalein blue, alpha-naphtholphtalein, 3,3-bis 4-hydroxy-1-naphthyl phthalide | |
| C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O | |
| 418.448 | |
| 418.45 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3350
TSCA : No