missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Lactose Monohydrate, Spectrum™ Chemical
L2071, 5989-81-1, C12H22O11·H2O
$1038.15 - $1038.15
Chemical Identifiers
| CAS | 5989-81-1 |
|---|---|
| Molecular Formula | C12H24O12 |
| Molecular Weight (g/mol) | 360.31 |
| InChI Key | WSVLPVUVIUVCRA-KPKNDVKVSA-N |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate |
| SMILES | O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O |
Description
Spectrum™ Chemical alpha-Lactose Monohydrate , also known as milk sugar, is a disaccharide sugar and is added to pills as a filler because of its physical properties. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 5989-81-1 | |
| 360.31 | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate |
| C12H24O12 | |
| WSVLPVUVIUVCRA-KPKNDVKVSA-N | |
| O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O |
Specifications
| 5989-81-1 | |
| Poly Pail | |
| 5 kg | |
| O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O | |
| 360.31 |
| 100% | |
| C12H24O12 | |
| WSVLPVUVIUVCRA-KPKNDVKVSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate | |
| Reagent |