missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-(+)-Talose, 97%

CAS: 2595-98-4 | C6H12O6 | 180.16 g/mol

Supplier:  Thermo Scientific Chemicals B2122677

 View more versions of this product

Catalog No. AAB2122677


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

alpha-D-(+)-Talose
133°C to 135°C
MFCD00135834
d-+-talose, aldehydo-d-talose, 2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal
OCC(O)C(O)C(O)C(O)C=O
180.16
CHEBI:68461
97%
2595-98-4
C6H12O6
0.1 g
GZCGUPFRVQAUEE-UHFFFAOYNA-N
2,3,4,5,6-pentahydroxyhexanal
99459
180.16

Chemical Identifiers

2595-98-4
180.16
GZCGUPFRVQAUEE-UHFFFAOYNA-N
99459
2,3,4,5,6-pentahydroxyhexanal
C6H12O6
MFCD00135834
d-+-talose, aldehydo-d-talose, 2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal
CHEBI:68461
OCC(O)C(O)C(O)C(O)C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-996-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only