CAS RN 2595-98-4
IUPAC Name:
2,3,4,5,6-pentahydroxyhexanal
Synonyms:
dextrose corn sugar (14C)-glucose D(+)-glucose (U-14C)glucose (14C)D-glucose sirup D-(U-14C)glucose glucose (D) D(+)-mannose
Molecular Weight (g/mol):
180.16
Molecular Formula:
C6H12O6
InChi Key:
GZCGUPFRVQAUEE-UHFFFAOYNA-N
SMILES:
OCC(O)C(O)C(O)C(O)C=O
1
–
2
of
2
results
1
Promotions Available
alpha-D-(+)-Talose, 97%
alpha-D-(+)-Talose, 97%
CAS: 2595-98-4 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 MDL Number: MFCD00135834 InChI Key: GZCGUPFRVQAUEE-UHFFFAOYNA-N Synonym: d-+-talose,aldehydo-d-talose,2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal PubChem CID: 99459 ChEBI: CHEBI:68461 IUPAC Name: 2,3,4,5,6-pentahydroxyhexanal SMILES: OCC(O)C(O)C(O)C(O)C=O
2
D-(+)-Talose 98.0+%, TCI America™
CAS: 2595-98-4 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 MDL Number: MFCD00135834 InChI Key: GZCGUPFRVQAUEE-UHFFFAOYNA-N Synonym: d-+-talose,aldehydo-d-talose,2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal PubChem CID: 99459 ChEBI: CHEBI:68461 IUPAC Name: 2,3,4,5,6-pentahydroxyhexanal SMILES: OCC(O)C(O)C(O)C(O)C=O