missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloro-o-xylene 98.0+%, TCI America™

Catalog No. C086425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C086425G 25 g
C0864500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C086425G Supplier TCI America Supplier No. C086425G

Chemical Identifiers

CAS 552-45-4
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000899
InChI Key VQRBXYBBGHOGFT-UHFFFAOYSA-N
Synonym 2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw
PubChem CID 11093
IUPAC Name 1-(chloromethyl)-2-methylbenzene
SMILES CC1=CC=CC=C1CCl

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 g
UN Number 3265
Formula Weight 140.61
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Chloro-o-xylene
EINECSNumber (3)-2691
TSCA Yes