accessibility menu, dialog, popup

UserName

CH3Cl

CAS RN 552-45-4

IUPAC Name: 1-(chloromethyl)-2-methylbenzene
Synonyms: 1-(chloromethyl)-2-methylbenzene
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: VQRBXYBBGHOGFT-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CCl
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)