missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromodiphenylmethane 97.0+%, TCI America™

Catalog No. B058625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B058625G 25 g
B0586100G 100 g
B0586500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B058625G Supplier TCI America Supplier No. B058625G

Chemical Identifiers

CAS 776-74-9
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.135
MDL Number MFCD00000134
InChI Key OQROAIRCEOBYJA-UHFFFAOYSA-N
Synonym bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane
PubChem CID 236603
IUPAC Name [bromo(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Br

Specifications

Melting Point 39°C
Boiling Point 193°C
Quantity 25 g
UN Number 1770
Formula Weight 247.14
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material alpha-Bromodiphenylmethane
RTECSNumber PA5350000
TSCA Yes
Recommended Storage Refrigerator