missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-o-xylene 97.0+%, TCI America™

Catalog No. p-7118798
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B100425G 25 g
B1004100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B100425G Supplier TCI America Supplier No. B100425G

Chemical Identifiers

CAS 89-92-9
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000174
InChI Key WGVYCXYGPNNUQA-UHFFFAOYSA-N
Synonym 2-methylbenzyl bromide, 1-bromomethyl-2-methylbenzene, alpha-bromo-o-xylene, o-xylyl bromide, o-methylbenzyl bromide, 2-xylyl bromide, 2-bromomethyl toluene, 2-methylbenzylbromide, benzene, 1-bromomethyl-2-methyl, a-bromo-o-xylene
PubChem CID 6992
IUPAC Name 1-(bromomethyl)-2-methylbenzene
SMILES CC1=CC=CC=C1CBr

Specifications

Color Yellow
Boiling Point 224°C
Quantity 25 g
UN Number 1701
Formula Weight 185.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Bromo-o-xylene