accessibility menu, dialog, popup

UserName

CH3Br

CAS RN 89-92-9

IUPAC Name: 1-(bromomethyl)-2-methylbenzene
Synonyms: 1-(bromomethyl)-2-methylbenzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: WGVYCXYGPNNUQA-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CBr
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)