missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Bromo-4-chlorophenylacetic Acid 98.0+%, TCI America™
$95.06 - $277.75
Chemical Identifiers
| CAS | 3381-73-5 |
|---|---|
| Molecular Formula | C8H6BrClO2 |
| Molecular Weight (g/mol) | 249.49 |
| MDL Number | MFCD08276760 |
| InChI Key | KKOAAWLOOHBFQP-UHFFFAOYNA-N |
| Synonym | 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid |
| PubChem CID | 10490868 |
| IUPAC Name | 2-bromo-2-(4-chlorophenyl)acetic acid |
| SMILES | OC(=O)C(Br)C1=CC=C(Cl)C=C1 |
Chemical Identifiers
| 3381-73-5 | |
| 249.49 | |
| KKOAAWLOOHBFQP-UHFFFAOYNA-N | |
| 10490868 | |
| OC(=O)C(Br)C1=CC=C(Cl)C=C1 |
| C8H6BrClO2 | |
| MFCD08276760 | |
| 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid | |
| 2-bromo-2-(4-chlorophenyl)acetic acid |
Specifications
| 98°C | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| MFCD08276760 | |
| 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid | |
| OC(=O)C(Br)C1=CC=C(Cl)C=C1 | |
| 249.49 | |
| 249.49 | |
| alpha-Bromo-4-chlorophenylacetic Acid |
| 3381-73-5 | |
| 5 g | |
| C8H6BrClO2 | |
| 3261 | |
| KKOAAWLOOHBFQP-UHFFFAOYNA-N | |
| 2-bromo-2-(4-chlorophenyl)acetic acid | |
| 10490868 | |
| ≥98.0% (T) |
Safety and Handling
TSCA : No