missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Bromo-4-chlorophenylacetic Acid 98.0+%, TCI America™
Supplier: TCI America B336725G
Specifications
| alpha-Bromo-4-chlorophenylacetic Acid | |
| 3381-73-5 | |
| 25 g | |
| C8H6BrClO2 | |
| 3261 | |
| KKOAAWLOOHBFQP-UHFFFAOYNA-N | |
| 2-bromo-2-(4-chlorophenyl)acetic acid | |
| 10490868 | |
| ≥98.0% (T) |
| 98°C | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| MFCD08276760 | |
| 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid | |
| OC(=O)C(Br)C1=CC=C(Cl)C=C1 | |
| 249.49 | |
| 249.49 |
Chemical Identifiers
| 3381-73-5 | |
| 249.49 | |
| KKOAAWLOOHBFQP-UHFFFAOYNA-N | |
| 10490868 | |
| OC(=O)C(Br)C1=CC=C(Cl)C=C1 |
| C8H6BrClO2 | |
| MFCD08276760 | |
| 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid | |
| 2-bromo-2-(4-chlorophenyl)acetic acid |
Safety and Handling
TSCA : No