accessibility menu, dialog, popup

UserName

OOHBrCl

CAS RN 3381-73-5

IUPAC Name: 2-bromo-2-(4-chlorophenyl)acetic acid
Synonyms: bromo(4-chlorophenyl)acetic acid
Molecular Weight (g/mol): 249.49
Molecular Formula: C8H6BrClO2
InChi Key: KKOAAWLOOHBFQP-UHFFFAOYNA-N
SMILES: OC(=O)C(Br)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)