Learn More
alpha,alpha-Dimethylbenzylamine, 96%
CAS: 585-32-0 | C9H13N | 135.21 g/mol
$370.71 - $1099.78
Chemical Identifiers
| CAS | 585-32-0 |
|---|---|
| Molecular Formula | C9H13N |
| Molecular Weight (g/mol) | 135.21 |
| MDL Number | MFCD00134680 |
| InChI Key | KDFDOINBXBEOLZ-UHFFFAOYSA-N |
| Synonym | cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine |
| PubChem CID | 68509 |
| IUPAC Name | 2-phenylpropan-2-amine |
| SMILES | CC(C)(N)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2684503
|
Thermo Scientific Chemicals
H2684503 |
1 g |
Each for $370.71
|
|
|||||
|
AAH2684506
|
Thermo Scientific Chemicals
H2684506 |
5 g |
Each for $1,099.78
|
|
|||||
Description
Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAgrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Chemical Identifiers
| 585-32-0 | |
| 135.21 | |
| KDFDOINBXBEOLZ-UHFFFAOYSA-N | |
| 68509 | |
| CC(C)(N)C1=CC=CC=C1 |
| C9H13N | |
| MFCD00134680 | |
| cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine | |
| 2-phenylpropan-2-amine |
Specifications
| 585-32-0 | |
| 197°C | |
| Amine-like | |
| 1.52 | |
| 1 g | |
| Air Sensitive | |
| Insoluble in water. | |
| CC(C)(N)C1=CC=CC=C1 | |
| 135.21 | |
| 135.21 | |
| alpha,alpha-Dimethylbenzylamine |
| 0.94 | |
| 72°C (162°F) | |
| C9H13N | |
| MFCD00134680 | |
| UN2735 | |
| cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine | |
| KDFDOINBXBEOLZ-UHFFFAOYSA-N | |
| 2-phenylpropan-2-amine | |
| 68509 | |
| 96% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
RUO – Research Use Only