missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dimethylbenzylamine, 96%

Catalog No. AAH2684503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2684503 1 g
AAH2684506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2684503 Supplier Thermo Scientific Chemicals Supplier No. H2684503

CAS: 585-32-0 | C9H13N | 135.21 g/mol

Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 585-32-0
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00134680
InChI Key KDFDOINBXBEOLZ-UHFFFAOYSA-N
Synonym cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine
PubChem CID 68509
IUPAC Name 2-phenylpropan-2-amine
SMILES CC(C)(N)C1=CC=CC=C1

Specifications

Density 0.94
Boiling Point 197°C
Flash Point 72°C (162°F)
Odor Amine-like
Refractive Index 1.52
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 135.21
Percent Purity 96%
Chemical Name or Material alpha,alpha-Dimethylbenzylamine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No

RUO – Research Use Only