missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha'-Dihydroxy-1,4-diisopropylbenzene 97.0+%, TCI America™
Supplier: TCI America D157725G
Specifications
| alpha,alpha′-Dihydroxy-1,4-diisopropylbenzene | |
| 144°C | |
| 168°C | |
| MFCD00009827 | |
| 1,4-bis 2-hydroxy-2-propyl benzene, 1,4-bis 1-methyl-1-hydroxyethyl benzene, alpha,alpha,alpha',alpha'-tetramethyl-1,4-benzenedimethanol, unii-g785drb5xh, p-bis 2-hydroxyisopropyl benzene, 1,4-bis 2-hydroxyisopropyl benzene, p-bis alpha-hydroxyisopropyl benzene, g785drb5xh, alpha,alpha'-dihydroxy-p-diisopropylbenzene, alpha,alpha,alpha',alpha'-tetramethyl-p-xylylenediol | |
| CC(C)(C1=CC=C(C=C1)C(C)(C)O)O | |
| 194.274 | |
| 194.27 | |
| Crystalline Powder |
| 2948-46-1 | |
| White-Yellow | |
| C12H18O2 | |
| 25 g | |
| LEARFTRDZQQTDN-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol | |
| 18062 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 2948-46-1 | |
| 194.274 | |
| LEARFTRDZQQTDN-UHFFFAOYSA-N | |
| 18062 | |
| CC(C)(C1=CC=C(C=C1)C(C)(C)O)O |
| C12H18O2 | |
| MFCD00009827 | |
| 1,4-bis 2-hydroxy-2-propyl benzene, 1,4-bis 1-methyl-1-hydroxyethyl benzene, alpha,alpha,alpha',alpha'-tetramethyl-1,4-benzenedimethanol, unii-g785drb5xh, p-bis 2-hydroxyisopropyl benzene, 1,4-bis 2-hydroxyisopropyl benzene, p-bis alpha-hydroxyisopropyl benzene, g785drb5xh, alpha,alpha'-dihydroxy-p-diisopropylbenzene, alpha,alpha,alpha',alpha'-tetramethyl-p-xylylenediol | |
| 2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol |
Safety and Handling
EINECSNumber : (3)-3620
TSCA : Yes