accessibility menu, dialog, popup

UserName

CH3H3COHCH3H3COH

CAS RN 2948-46-1

IUPAC Name: 2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol
Synonyms: 2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: LEARFTRDZQQTDN-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=CC=C(C=C1)C(C)(C)O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)