missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dihydroxy-1,4-diisopropylbenzene 97.0+%, TCI America™

Catalog No. D15775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15775G 5 g
D157725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15775G Supplier TCI America Supplier No. D15775G

Chemical Identifiers

CAS 2948-46-1
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.274
MDL Number MFCD00009827
InChI Key LEARFTRDZQQTDN-UHFFFAOYSA-N
Synonym 1,4-bis 2-hydroxy-2-propyl benzene, 1,4-bis 1-methyl-1-hydroxyethyl benzene, alpha,alpha,alpha',alpha'-tetramethyl-1,4-benzenedimethanol, unii-g785drb5xh, p-bis 2-hydroxyisopropyl benzene, 1,4-bis 2-hydroxyisopropyl benzene, p-bis alpha-hydroxyisopropyl benzene, g785drb5xh, alpha,alpha'-dihydroxy-p-diisopropylbenzene, alpha,alpha,alpha',alpha'-tetramethyl-p-xylylenediol
PubChem CID 18062
IUPAC Name 2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES CC(C)(C1=CC=C(C=C1)C(C)(C)O)O

Specifications

Melting Point 144°C
Color White-Yellow
Boiling Point 168°C
Quantity 5 g
Formula Weight 194.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Dihydroxy-1,4-diisopropylbenzene
EINECSNumber (3)-3620
TSCA Yes