missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dichloro-p-xylene 98.0+%, TCI America™

Catalog No. D043025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D043025G 25 g
D0430100G 100 g
D0430500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D043025G Supplier TCI America Supplier No. D043025G

Chemical Identifiers

CAS 623-25-6
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000920
InChI Key ZZHIDJWUJRKHGX-UHFFFAOYSA-N
Synonym 1,4-bis chloromethyl benzene, alpha,alpha'-dichloro-p-xylene, p-xylylene dichloride, p-xylylene chloride, benzene, 1,4-bis chloromethyl, p-bis chloromethyl benzene, benzene, p-bis chloromethyl, a,a'-dichloro-p-xylene, xylylene dichloride, ccris 1772
PubChem CID 12171
IUPAC Name 1,4-bis(chloromethyl)benzene
SMILES C1=CC(=CC=C1CCl)CCl

Specifications

Melting Point 100°C
Boiling Point 142°C
Quantity 25 g
UN Number 2928
Formula Weight 175.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Dichloro-p-xylene
EINECSNumber (3)-0047
RTECSNumber ZE4100000
TSCA Yes