missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha'-Dichloro-m-xylene 96.0+%, TCI America™
$104.38 - $859.33
Chemical Identifiers
| CAS | 626-16-4 |
|---|---|
| Molecular Formula | C8H8Cl2 |
| Molecular Weight (g/mol) | 175.052 |
| MDL Number | MFCD00000912 |
| InChI Key | GRJWOKACBGZOKT-UHFFFAOYSA-N |
| Synonym | 1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro |
| PubChem CID | 12275 |
| IUPAC Name | 1,3-bis(chloromethyl)benzene |
| SMILES | C1=CC(=CC(=C1)CCl)CCl |
Chemical Identifiers
| 626-16-4 | |
| 175.052 | |
| GRJWOKACBGZOKT-UHFFFAOYSA-N | |
| 12275 | |
| C1=CC(=CC(=C1)CCl)CCl |
| C8H8Cl2 | |
| MFCD00000912 | |
| 1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro | |
| 1,3-bis(chloromethyl)benzene |
Specifications
| 626-16-4 | |
| C8H8Cl2 | |
| 25 g | |
| 1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro | |
| C1=CC(=CC(=C1)CCl)CCl | |
| 175.052 | |
| 175.05 | |
| Crystalline Lumps |
| White | |
| MFCD00000912 | |
| 2928 | |
| GRJWOKACBGZOKT-UHFFFAOYSA-N | |
| 1,3-bis(chloromethyl)benzene | |
| 12275 | |
| ≥96.0% (GC) | |
| alpha,alpha′-Dichloro-m-xylene |
Safety and Handling
EINECSNumber : (3)-0047&(9)-1869
RTECSNumber : ZE4070000
TSCA : Yes