accessibility menu, dialog, popup

UserName

CAS RN 626-16-4

IUPAC Name: 1,3-bis(chloromethyl)benzene
Synonyms: 1,3-(bis-chloromethyl)benzene
Molecular Weight (g/mol): 175.05
Molecular Formula: C8H8Cl2
InChi Key: GRJWOKACBGZOKT-UHFFFAOYSA-N
SMILES: ClCC1=CC(CCl)=CC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)