missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha'-Dichloro-m-xylene 96.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| UN Number | 2928 |
| Formula Weight | 175.05 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | alpha,alpha′-Dichloro-m-xylene |
Safety and Handling
| EINECSNumber | (3)-0047&(9)-1869 |
| RTECSNumber | ZE4070000 |
| TSCA | Yes |